Learn More
Sino Biological Anti-Prolactin Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10278T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Prolactin Receptor/PRLR (rh Prolactin Receptor/PRLR; Catalog#10278-H08H; NP_000940.1; Met1-Asp234). Prolactin Receptor/PRLR specific IgG was purified by Human Prolactin Receptor/PRLR affinity chromatography.
Catalog No. 50-172-3987