Learn More
Sino Biological Anti-C11orf57 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102848T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human C11orf57, and purified by antigen affinity chromatography.
Catalog No. 50-172-4044