Learn More
Sino Biological Coagulation Factor XI / FXI / F11 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10302RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Factor XI (rh Factor XI; Catalog#10302-H08H; NP_000119.1; Met 1-Val 625). Factor XI specific IgG was purified by human Factor XI affinity chromatography.
Catalog No. 50-172-4260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.