Learn More
Sino Biological Anti-Osteopontin/OPN Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10352RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SPP1 (rh SPP1; Catalog#10352-H08H; NP_001035147.1; Met 1-Asn 314). SPP1 specific IgG was purified by human SPP1 affinity chromatography.
Catalog No. 50-172-4903