Learn More
Epredia™ Cytology Funnel Clips
Catalog No. 10357 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Metal
Catalog No. Material
10-357 Metal
1 options
This item is not returnable. View return policy
Catalog No. 10-357
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
For use with Shandon™ and Fisherbrand™ cytology funnels
  • Functionally identical to the Shandon Cytoclip
  • Compatible with Fisherbrand Cytology Funnels
  • Compatible with Fisherbrand Ringed Cytology Slides
  • All-metal construction
Specifications
Type Cytology Funnel Clip
Material Metal
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov