Learn More
Sino Biological Recombinant Human Thioredoxin/TRX Protein, HPLC-verified
Supplier:  Sino Biological 10384HNAE500
Encompass_Preferred
A DNA sequence encoding the C-terminal segment of human TXN (P10599) (Met 1-Val 105) was expressed and purified.
Catalog No. 50-161-4811
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.