Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-[3-(Trimethoxysilyl)Propyl]Ethylenediamine

Catalog No. 104884500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-488-4500ML 500 mL
10-488-4100ML 100 mL
10-488-41L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 10-488-4500ML Supplier Sigma Aldrich Supplier No. 104884500ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-[3-(Trimethoxysilyl)propyl]ethylenediamine has also been used as an organic ligand for the surface modification of silica gel to uptake heavy metal ions. It can react with fluorinated carbon nanotubes (F-CNT) to form aminoalkylalkoxysilane functionalized carbon nanotubes.

Specifications

CAS 1760-24-3
Density 1.028 g/mL at 25°C
Boiling Point 146°C (15 mmHg, lit.)
Molecular Formula C8H22N2O3Si
Refractive Index n20/D 1.444
Linear Formula (CH3O)3Si(CH2)3NHCH2CH2NH2
MDL Number MFCD00008173
Quantity 500 mL
Synonym [3-(2-Aminoethylamino)propyl]trimethoxysilane; N-(2-Aminoethyl)-3-(trimethoxysilyl)propylamine
Molecular Weight (g/mol) 222.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.