Learn More
Sino Biological Human IL1F6 / IL36A Protein (His Tag) 20ug
Supplier:  Sino Biological 10607H07E120
Encompass_Preferred
A DNA sequence encoding the mature form of human IL1F6 (Q9UHA7) (Lys 6-Phe158) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5287
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.