Learn More
Sino Biological Human CCL23 / MIP 3 Protein 50ug
Supplier:  Sino Biological 10655HNCE50
Encompass_Preferred
A DNA sequence encoding the human CCL23 (AAB51134.1) (Arg22-Asn120) was expressed.
Catalog No. 50-161-5344
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.