Learn More
Sino Biological Human Beta-amyloid 38 / Beta-APP38 Protein (aa 672-709, His & GST Tag) 100ug
Supplier:  Sino Biological 10703H20E4100
Encompass_Preferred
A DNA sequence encoding the amino acids (Asp672-Gly709) of human Amyloid beta A4 protein (APP770) (P05067-1), corresponding to the Beta-amyloid protein 38, was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-5476
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.