Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyruvic acid

Catalog No. 107360100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-736-0100G 100 g
10-736-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-736-0100G Supplier Sigma Aldrich Supplier No. 107360100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pyruvic acid

Specifications

CAS 127-17-3
Melting Point 11°C to 12°C (lit.)
Density 1.267 g/mL (at 25°C (literature))
Boiling Point 165°C (lit.)
Molecular Formula C3H4O3
Linear Formula CH3COCOOH
Refractive Index n20/D 1.428 (literature)
MDL Number MFCD00002585
Quantity 100 g
Synonym alpha-Ketopropionic acid; 2-Oxopropionic acid
Molecular Weight (g/mol) 88.06
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.