Learn More
Sino Biological Recombinant Human VEGFR3/FLT4 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10806H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ile 776) of human VEGFR3 (NP_002011.2) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.