Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexanecarboxaldehyde

Catalog No. 10846425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-846-425G 25 g
10-846-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-846-425G Supplier Sigma Aldrich Supplier No. 10846425G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cyclohexanecarboxaldehyde was used to study gas-phase microwave spectrum of cyclohexanecarboxaldehyde
  • It was used for the synthesis of poly(vinyl cyclohexanal)
  • It was used as model substrate in the synthesis of propargylamines using fe3o4 nanoparticles as catalyst
  • It was employed in an aldol reaction with ethyl diazoacetate catalyzed by basic, nanoparticulate magnesium oxide leading to α-diazo-α-hydroxyesters

Specifications

CAS 2043-61-0
Density 0.926 g/mL (at 25°C (literature))
Boiling Point 161°C to 163°C (lit.)
Molecular Formula C7H12O
Refractive Index n20/D 1.452 (literature)
Linear Formula C6H11CHO
MDL Number MFCD00001457
Quantity 25 g
Synonym Hexahydrobenzaldehyde
Molecular Weight (g/mol) 112.17
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.