Learn More
Selleck Chemical LLC Pyridostatin Trifluoroacetate Salt 5mg 1472611-44-1
SDP
Supplier:  Selleck Chemical LLC S74445MG
Encompass_Preferred
Pyridostatin Trifluoroacetate Salt is a G-quadruplexe stabilizer with Kd of 490 nM in a cell-free assay, which targets a series of proto-oncogenes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.