Learn More
Nusep Inc 10CM X 8.5CM EMPTY GEL CASSETT
Supplier:  Nusep Inc NB010
Encompass_Preferred
NuSep's 10cm x 8.5cm Empty Gel Cassette is designed to provide a reliable and reusable solution for custom gel preparation. Ideal for researchers who prefer to pour their own gels, this cassette supports various electrophoresis applications while maintaining durability and ease of use.Specifications:Dimensions: 10cm x 8.5cmUse: Suitable for custom gel casting and electrophoresisMaterial: Constructed for durability and repeated useNuSep’s empty gel cassette offers flexibility and consistency for laboratories looking to streamline their gel preparation process.
Catalog No. NC2457302
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.