Learn More
Postnova Analytics AF2000 MF - AF4 Analytic RC Membrane
SDP
Supplier:  Postnova Analytics ZAF4MEM61210KD
Encompass
10 kDa Regenerated Cellulose Membrane; Aqu/Org; 10-70°C; Size: 294 mm x 30 mm; 10 Pcs
Catalog No. NC1675412
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.