Learn More
Teknova 10mM Tris, 2mM Magnesium Chloride, 4% Sucrose, pH 8.0. 500mL. Sterile. 8 pack.
SDP
Supplier:  Teknova T16998PK
Encompass_Preferred
10mM Tris, 2mM Magnesium Chloride,4% Sucrose
Catalog No. 50-238-7619
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.