Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 10nm Colloidal Gold Nanoparticles 10OD
SDP

Catalog No. 30100020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-100-020 20 mL
30-100-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-100-020 Supplier Novus Biologicals™ Supplier No. 30100020

10nm colloidal gold nanoparticles ultra high quality, with narrow size distribution and uniform shape

  • 10nm colloidal gold nanoparticles are ultra high quality, with narrow size distribution and uniform shape, supplied in range of sizes and concentrations for maximum flexibility

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 20 mL
Kit Contents One 20mL bottle at 10 OD
Product Type 10nm Colloidal Gold Nanoparticles 10OD Antibody Labeling Kit

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.