Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 10nm Colloidal Gold Nanoparticles 10OD
SDP

Catalog No. 30100100 Shop All Novus Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-100-100 100 mL
30-100-020 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-100-100 Supplier R&D Systems™ Supplier No. 30100100

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. 10nm colloidal gold nanoparticles ultra high quality, with narrow size distribution and uniform shape

  • 10nm colloidal gold nanoparticles are ultra high quality, with narrow size distribution and uniform shape, supplied in range of sizes and concentrations for maximum flexibility

Specifications

Product Type 10nm Colloidal Gold Nanoparticles 10OD Antibody Labeling Kit
Content And Storage One 20mL bottle at 10 OD. Store at 4°C. Do not freeze.
Product Line Novus
Quantity 100 mL

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.