Description
- 10nm colloidal gold nanoparticles are ultra high quality, with narrow size distribution and uniform shape, supplied in range of sizes and concentrations for maximum flexibility
Specifications
Specifications
| Product Type | 10nm Colloidal Gold Nanoparticles 1OD Antibody Labeling Kit |
| Content And Storage | One 20mL bottle at 1 OD. Store at 4°C. Do not freeze. |
| Product Line | Novus |
| Quantity | 100 mL |
For research use only.