Learn More
Thomas Scientific Classic Low Retention Pipet Tips, 10 uL Racked Tip, Sterile, 960/pk
SDP
Supplier:  Thomas Scientific 1158U43
Encompass
These tips are the latest in low retention polymer technology and come to an extra fine point at the dispensing orifice, minimizing inaccuracies caused by surface adhesion and droplet retention. Their universal fit sealing ring design forms a superior seal on a broad range of pipette brands, including hard-to-fit P-20 pipettes.
Catalog No. NC1960102