Learn More
Chemglass Life Sciences Chemglass 10 μL Microliter Syringe, Model 701 N, 26s Gauge Cemented Needle, 2 in
SDP
Supplier:  Chemglass Life Sciences CG3072H03
Encompass_Preferred
Chemglass CG-3072-H03 (Model 701 N) is a 10 μL microliter syringe designed for precise micro-volume liquid delivery in analytical and laboratory applications. It features a glass barrel with a fitted plunger and a cemented needle for low dead volume transfers.
  • Precise 10 μL capacity for micro-volume dispensing.
  • Cemented 26s gauge needle for minimal dead volume.
  • 2 in needle length suitable for standard laboratory access.
  • Glass barrel with tight plunger fit for consistent performance.
  • Suitable for analytical sampling, microinjection, and micro-volume dosing.
Catalog No. NC2753192