Learn More
Bioassay Systems Polyvinylpyrrolidone (PVPP) used for preparation of wine samples for use with the QuantiQuik™ Histamine Quick Test Strips.
SDP
Catalog No. 501956333 Shop All Bioassay Systems Products
Encompass
Polyvinylpyrrolidone (PVPP) used for preparation of wine samples for use with the QuantiQuik™ Histamine Quick Test Strips.
Catalog No. 50-195-6333 Supplier Bioassay Systems Supplier No. PVPP10
Polyvinylpyrrolidone (PVPP) used for preparation of wine samples for use with the QuantiQuik™ Histamine Quick Test Strips.