Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 10X Dilution Buffer, 50 mL

Catalog No. 502587576
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-7576 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-7576 Supplier Abcam Supplier No. AB22228750ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Concentrated buffer solution designed for assay dilution applications.

10X Dilution Buffer is designed for use in assay kits.

Specifications

Form Liquid
Kit Contents 10X Dilution Buffer, 50 mL
Product Type Blocker and Diluent
Concentration 10X
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.