Learn More
Abcam 10x EDTA Buffer, pH 8.0, Antigen Retrieval, 125 mL
Description
- Applications: IHC-P
- Buffer constituents: 0.29% EDTA
- pH range: 7.7 to 8.1
- Form: Liquid
- Application notes: Heat-induced antigen retrieval (HIER) during IHC
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Format | Bottle |
| Form | Liquid |
| Buffer | 0.29% EDTA, pH7.7 to pH8.1 |
| Content And Storage | Store at 4°C. |
| Product Type | Wash Solution |
| Concentration | 10x |
| Formulation | 0.29% EDTA, pH 7.7 to 8.1 |
| Sample Type | Formalin-fixed paraffin-embedded tissue |
| Quantity | 125 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.