Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10X PBD, MP Biomedicals™

Catalog No. MP11RIST137 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP11RIST137 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP11RIST137 Supplier MP Biomedicals, Inc Supplier No. 11RIST1370
Add to Cart
Add to Cart

10X PBD

  • 10X Phosphate buffered detergent (PBD) used in enzymatic or fluorescence in situ hybridization (FISH). Intended for use with paraffin embedded, formalin-fixed tissue sections, whole cells interphase nuclei, and metaphase chromosomes.
  • 10X Phosphate-Buffered Detergent.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Physical Form Solution
Quantity 500 mL
Formulation SSC 4X and 0.5% TWIN 20
Synonym 10X Phosphate-Buffered Detergent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.