Learn More
Teknova 10X PBS. 1000mL, Sterile. Endotoxin Tested. 8 pack.
SDP
Supplier:  Teknova P77108PK
Encompass_Preferred
36.2mM Sodium Phosphate Monobasic,163.8mM Sodium Phosphate Dibasic,1.5M Sodium Chloride
Catalog No. 50-238-7505
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.