Learn More
Description
- Store in a dry, cool and well-ventilated place.
- Keep container closed when not in use. Keep/Store away from direct sunlight, extremely high or low temperatures and incompatible materials.
- Ensure adequate ventilation, especially in confined areas.
- Wear chemically resistant protective gloves.
Specifications
Specifications
| pH | 7.4 |
| Product Line | R&D Systems |
| Quantity | 500mL |
| Formulation | 75mM disodium hydrogen phosphate (Na2HPO4), 25mM sodium dihydrogen phosphate (NaH2PO4), 1.45M sodium chloride (NaCl) |
| Grade | Not Available |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
