Learn More
Teknova 10X PBS Modified Formula, 1000mL, Sterile 8 pack.
SDP
Supplier:  Teknova P02328PK
Encompass_Preferred
161.8mM Dipotassium Phosphate,38.2mM Monopotassium Phosphate,1.37M Sodium Chloride, 27mM Potassium Chloride
Catalog No. 50-238-7579
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.