Learn More
IBI SCIENTIFIC 3P 10X PBST (PBS W/0.5% TWEEN 20) 6/PACK
Supplier:  IBI SCIENTIFIC 3P IB701676
Encompass_Preferred
"Sterile buffer solution containing sodium phosphate, sodium chloride, potassium phosphate and potassium chloride with 0.5% TWEEN 20.Sterile filtered using 0.1um filterDNase, RNase and protease free.Sterility validated using USP <71> Test for SterilityDoes not contain calcium, magnesium, phenol red or sodium pyruvate. IBI 10X PBST (10X PBS) with TWEEN 20 (0.5% TWEEN 20) is a concentrated sterile solution in a 1 liter bottle that is ideal for use in immunoassay procedures such as Western Blotting and ELISAs in microtiter plate wells.IBI 10X PBST is mixed and bottled in our liquid production facility here in Dubuque, Iowa. Our high output pure water system produces endotoxin free and nuclease free water at a rate of 100 gallons per/hr. This water is the basis for all of our liquid blending production here at IBI."
Catalog No. NC2828990
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.