Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ T4 DNA Ligase Buffer (10X)

Catalog No. FERB69 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERB69 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERB69 Supplier Thermo Scientific™ Supplier No. B69
  • $42.75
    $35.65 / Each of 1

    Save $7.10

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Safely ligate samples with this buffer, for use with T4 DNA Ligase and tested for the absence of endo- and exodeoxyribonucleases and ribonucleases.

10X T4 DNA Ligase buffer is used with T4 DNA Ligase.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Quantity 1.5 mL
Product Type T4 DNA Ligase Buffer
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.