Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 10X TdT Stop Buffer
SDP

Catalog No. 48176003
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-176-003 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-176-003 Supplier R&D Systems™ Supplier No. 48176003

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ 10X TdT Stop Buffer is provided with the TACS™ TdT kits to stop the activity of the TdT enzyme in TUNEL labeling assays.

  • Long Name: Terminal Deoxynucleotidyl Transferase
  • Do not use past expiration date
  • There are no hazardous ingredients as defined in the United States 29 CFR 1910.1200 (OSHA) or the Canadian Controlled Products Regulations (WHMIS) and therefore not subject to the labelling requirements of the aforementioned standards.
  • Exposure may aggravate those with pre-existing eye, skin, or respiratory conditions
  • Store in a dry, cool, and well-ventilated place
  • Keep container closed when not in use
  • Keep/Store away from direct sunlight, extremely high or low temperatures, and incompatible materials
  • Uncontrolled product according to WHMIS classification criteria

Specifications

Quantity 20 mL
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.