Learn More
Bioassay Systems QuantiQuik™ Oxalate Quick Test Strips. Direct determination of oxalate concentrations in biological samples (e.g. serum, plasma, urine, etc.).
SDP
Catalog No. 501956346 Shop All Bioassay Systems Products
Encompass_Preferred
For rapid, direct determination of oxalate concentrations in biological samples. For Research Use Only (not for medical decisions). Key Features: Fast and sensitive. Use of 10 µL sample. Semi-quantitative measurement between 0-1.5 mg/dL or 0-170 µM (undiluted) Oxalate. Convenient. No expensive plate or cuvette readers required. Sample trea™ent and assay can be performed in under 10 minutes. Method: Visual. Samples: Serum, plasma, urine, fruits or vegetables, etc. Species: All. Procedure: Assay takes approximately 10 min. Kit size: 10 tests.
Catalog No. 50-195-6346 Supplier Bioassay Systems Supplier No. QQOXLT10
For rapid, direct determination of oxalate concentrations in biological samples. For Research Use Only (not for medical decisions). Key Features: Fast and sensitive. Use of 10 µL sample. Semi-quantitative measurement between 0-1.5 mg/dL or 0-170 µM (undiluted) Oxalate. Convenient. No expensive plate or cuvette readers required. Sample trea™ent and assay can be performed in under 10 minutes. Method: Visual. Samples: Serum, plasma, urine, fruits or vegetables, etc. Species: All. Procedure: Assay takes approximately 10 min. Kit size: 10 tests.