Learn More
AdipoGen 11-Bromo-1-undecanol
SDP
Supplier:  AdipoGen CDXB0626G050
Encompass_Preferred
Chemical. CAS 1611-56-9. Formula C11H23BrO. MW 251.2. 11-Bromo-1-undecanol is a bulding block for synthesis. It was used in the preparation of graft copolymers based on a metathesis skeleton with polymethyl methacrylate grafts and in the synthesis of polymerizable tail groups consisting of n-Alkoxy-2,4-hexadiene used in the design of radically polymerized ionic liquid crystal monomers.
Catalog No. 50-256-1577
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.