Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11-Bromo-1-undecene 94.0+%, TCI America™
SDP

Catalog No. B357625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B357625G 25 g
B35765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B357625G Supplier TCI America Supplier No. B357625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7766-50-9
Molecular Formula C11H21Br
Molecular Weight (g/mol) 233.193
MDL Number MFCD00040825
InChI Key YPLVPFUSXYSHJD-UHFFFAOYSA-N
Synonym 11-bromo-1-undecene, 10-undecenyl bromide, 1-bromo-10-undecene, undec-10-enyl bromide, 11-bromo-undec-1-ene, 1-undecene, 11-bromo, 11-bromoundecene, undecylenyl bromide, n-undecylenic bromide, w-undecylenyl bromide
PubChem CID 284148
IUPAC Name 11-bromoundec-1-ene
SMILES C=CCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
Formula Weight 233.19
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 11-Bromo-1-undecene
EINECSNumber (2)-0067
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.