Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11-Bromoundecanoic Acid 99+%, Thermo Scientific™

Catalog No. AC107520250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC107520250 25 g
AC107521000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107520250 Supplier Thermo Scientific Chemicals Supplier No. 107520250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2834-05-1
Molecular Formula C11H21BrO2
Molecular Weight (g/mol) 265.19
InChI Key IUDGNRWYNOEIKF-UHFFFAOYSA-N
Synonym undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid
PubChem CID 17812
IUPAC Name 11-bromoundecanoic acid
SMILES C(CCCCCBr)CCCCC(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 48°C to 51°C
Color White to Beige
Infrared Spectrum Authentic
Linear Formula Br(CH2)10CO2H
Quantity 25 g
Beilstein 02, II, 315
Formula Weight 265.19
Percent Purity ≥99.0%
Physical Form Crystalline Flakes
Chemical Name or Material 11-Bromoundecanoic acid, 99+%
Show More Show Less
EINECSNumber 220-602-9
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.