Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 11-Dehydrothromboxane B2 Monoclonal Antibody (3A1)

Catalog No. PIMA549749
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549749 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549749 Supplier Invitrogen™ Supplier No. MA549749

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

11-Dehydrothromboxane B2 (11-DHTXB2) is a biomarker involved in the production of thromboxane, a molecule that contributes to platelet aggregation and vasoconstriction. It is produced by platelets, macrophages, and endothelial cells. Elevated levels of 11-DHTXB2 are associated with cardiovascular disease, stroke, and cancer. It is being investigated as a potential biomarker for predicting cardiovascular events and monitoring the effectiveness of antiplatelet therapies. 11-DHTXB2 may also have potential as a therapeutic target for certain types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 11-Dehydrothromboxane B2
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3A1
Concentration 11.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of 11-dhTxB2 hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.