Learn More
Medchemexpress LLC 11-mercaptoundecanoic acid | 71310-21-9 | MFCD00022096 | 99.8% | 218.36 g·mol⁻1 | C11H22O2S | 1 G

Supplier: Medchemexpress LLC HYW0140691G
11-mercaptoundecanoic acid is a thiol-terminated alkyl carboxylic acid commonly used to form self-assembled monolayers (SAMs) on metal surfaces and as a carboxyl-functional linker for surface modification, bioconjugation, and materials research.
- High purity 99.84% as supplied.
- Molecular weight 218.36 g·mol⁻¹, formula C11H22O2S.
- Solid, white to off-white appearance.
- Available in small solid packs and as a DMSO solution for immediate use.
- Powder stability at -20°C for long-term storage; stock solutions stable at -80°C.
- Effective for SAM formation on gold and for carboxyl-based surface coupling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.