Learn More
AdipoGen 11-O-Methylpseurotin A (250 µg)
SDP
Supplier:  AdipoGen AGCN20121C250
Encompass_Preferred
Chemical. CAS: 956904-34-0. Formula: C23H27NO8. MW: 445.5. Isolated from Sporothrix sp. Antibacterial. Selectively inhibits a Hof1 deletion strain.
Catalog No. 50-205-3488
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.