Learn More
Protocol™ Decalcifying Solution B, Fisherbrand™
SDP
Catalog No. 11002203 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-203 1 gal.
11-002-204 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-203
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Decalcifies the average bone marrow biopsy in less than one hour
  • Can be used on all types of calcified tissue and is especially effective on bone marrow samples
  • Cellular and nuclear details remain crisp and clear
Specifications
Product Type Decalcifying Solution
Quantity 1 gal.
Solution Type Decalcifier
Form Solution
Made in the USA