Learn More
Description
- 10% neutral buffered formalin fixative
- Used to preserve the cellular structure and morphology of tissue for examination under a microscope by preventing decay and maintaining its molecular integrity during the tissue processing steps for staining and analysis
- Featuring a pH level of 7.0
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Histological |
| Product Type | Formalin |
| Quantity | 2.5 gal. |
| Solution Type | Formalin |
| Form | Solution |
Made in the USA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
