Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocol™ Zinc Formalin, Fisherbrand™
SDP

Catalog No. 11002224 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-224 1 gal.
11-002-225 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-224
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Ready-to-use buffered zinc formalin in bulk packaging.

  • Especially recommended for use in immunohistochemistry, as zinc formalin preserves more immunoreactivity than standard NBF.
  • Used as a fixative solution that provides excellent preservation of tissue morphology, particularly nuclear and cytoplasmic components, by increasing the rigidity of cellular structures

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 1 gal.
Solution Type Formalin
Form Solution

Made in the USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.