Learn More
Protocol™ Define Clarifier, Fisherbrand™
SDP
Catalog No. 11002234 Shop All Fisherbrand Products
Change view
Click to view available options
1 gal.
Catalog No.
11-002-234 1 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-234
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Provides unparalleled crisp nuclear chromatin detail and eliminates any undesired background staining
  • A glacial acid based formulation.
  • Premixed, safe, and ready-to-use.
  • Gallon Size
Specifications
Product Type Xylene Substitute
Form Solution
Made in the USA