Learn More
Sino Biological Human ABL1 / JTK7 / p150 Protein (GST Tag) 50ug
Supplier:  Sino Biological 11199H09B50
Encompass_Preferred
A DNA sequence encoding the amino acid sequence (Pro 137-Ser 554) of human ABL1 isoform B (NP_009297.2) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-6661
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.