Learn More
Marathon LS 12 Well Tissue Culture Plate with Lid, Individual, Sterile 100 per case
SDP
Supplier:  Marathon LS MLSTCPRW12IL
Encompass_Preferred
12 Well Tissue Culture Plate with Lid, Individual, Sterile, 100 per case. Polystyrene. Flat Bottom with Lid. Tissue Culture Treated. Cell Growth Area: 3.85cm2. RNAse/DNase Free. Non-pyrogenic. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates.
Catalog No. NC2065687
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.