Learn More
Sino Biological Human AGO3 / Argonaute 3 / EIF2C3 Protein (His Tag) 20ug
Supplier:  Sino Biological 11433H07B20
Encompass_Preferred
A DNA sequence encoding the human EIF2C3 (Q9H9G7-1) (Met 1-Ala 860) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6888
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.