Learn More
Sino Biological Influenza A H11N2 (A/duck/Yangzhou/906/2002) Hemagglutinin Protein (HA1 Subunit) (His Tag) 100ug
Supplier:  Sino Biological 11705V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 342) of the influenza A hemagglutinin H11N2 (A/duck/Yangzhou/906/2002 (H11N2)) (AAY85533.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7291
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.