Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Diaminopropane

Catalog No. 117498100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-749-8100G 100 g
11-749-8500G 500 g
11-749-82KG 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-749-8100G Supplier Sigma Aldrich Supplier No. 117498100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,2-diaminopropane undergoes condensation with salicylaldehyde and its derivatives to form schiff base
  • It reacts with formaldehyde and hydrogen peroxide in aqueous acidic solution to give cage peroxide, 1,6-diaza-3,4,8,9-tetraoxa-11-methylbicyclo[4.4.2]dodecane.1,2-diaminopropane was used as model precursor in the electron induced deposition of amorphous carbon nitride films

Specifications

CAS 78-90-0
Density 0.87 g/mL (at 25°C (literature))
Boiling Point 119°C to 120°C (lit.)
Molecular Formula C3H10N2
Refractive Index n20/D 1.446 (literature)
Linear Formula CH3CH(NH2)CH2NH2
MDL Number MFCD00008089
Quantity 100 g
Synonym 1,2-Propanediamine; Propylenediamine
Molecular Weight (g/mol) 74.12
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.