Learn More
Sino Biological Human IL4 / Interleukin-4 Protein 1mg
Supplier:  Sino Biological 11846HNAE1
Encompass_Preferred
A DNA sequence encoding the mature form of human IL4 isoform 1 (P05112-1) (His 25-Ser 153) was expressed and purified, with an initial Met.
Catalog No. 50-161-7540
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.