Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ RPMI 1640 Medium
GREENER_CHOICE

Catalog No. 11875119
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
10 x 500 mL
20 x 100 mL
6 x 1000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-875-119 10 x 500 mL
11-875-101 100 mL
11-875-127 20 x 100 mL
11-875-093 500 mL
11-875-085 1000 mL
11-875-135 6 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 11-875-119 Supplier Gibco™ Supplier No. 11875119
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Requires

Supplementation

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer.

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 Medium modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI is modified as follows:

With: L-glutamine, Phenol Red

Without: HEPES

The complete formulation is available.

Using RPMI

RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell Type Leukemic Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Without Additives No HEPES, No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 10 x 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less
Do you have suggestions for improving viability when transfecting HL-60 cells with the Neon system?

Our online protocol suggests to use 50,000 cells with the 10 microliter tip. However, HL-60 cells do not survive well when transfected at low density. To improve viability you can try using at least 100,000 (better: 200,000) cells with the 10 microliter tips. Also, make sure your plasmid DNA is highly purified, as HL-60 cells are sensitive to LPS. If you observe that viability is good after 24 hours but decreases over the next 72 hours, you may either be using the wrong culture medium (RPMI 1640 + 10% FBS is recommended, do not use DMEM) or your batch of FBS contains low levels of cytotoxic contaminants.

Other possibilities are mycoplasma contamination or very high passage number of your cells. If this is the case please buy a fresh vial of HL-60 cells from ATCC and try your transfections again.

How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

What is the density (g/L) for RPMI 1640 Medium?

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

My cells are not adhering to the culture vessel. What should I do?

This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.

I see a precipitate in my medium, and a change in the pH. What should I do?

This is most likely a sign of bacterial or fungal contamination. We would recommend discarding the media, and trying to decontaminate the culture.

I see a precipitate in my medium, but no change in the pH. What should I do?

The precipitate could be due to residual phosphate left over from detergent washing, which may precipitate powered medium components. Rinse glassware in deionized, distilled water several times, then sterilize. If the medium is frozen, try warming media to 37 degrees C and swirl to dissolve. If precipitate remains, discard medium.

I'm seeing a rapid pH shift in my medium. What should I do?

Please see the possible causes for this rapid pH shift, and suggested solutions:

- Incorrect carbon dioxide tension: Increase or decrease percentage of carbon dioxide in the incubator based on concentration of sodium bicarbonate in the medium. For sodium bicarbonate concentrations of 2.0 to 3.7 g/L, use carbon dioxide amounts of 5-10%, respectively. Switch to carbon dioxide-independent medium.
- Overly tight caps on tissue culture flasks: Loosen caps one-quarter turn.
- Insufficient bicarbonate buffering: Add HEPES buffer to a final concentration of 10-25 mM.
- Incorrect salts in medium: Use an Earle's salts-based medium in a CO2 environment and a Hanks' salts-based medium in atmospheric conditions.
- Bacterial, yeast, or fungal contamination: Discard culture and medium. Try to decontaminate culture.

My cells are growing very slowly. What could be the cause of this?

Please see some common reasons below, and solutions to the issue:

- Growth medium is not correct: Use pre-warmed growth medium as recommended by the supplier.
- Serum in the growth medium is of poor quality: Use serum from a different lot.
- Cells have been passaged too many times: Use healthy, low-passage number cells.
- Cells were allowed to grow beyond confluency: Passage mammalian cells when they are in the log-phase before they reach confluency.
- Culture is contaminated with mycoplasma: Discard cells, media, and reagents. Obtain new stocks of cells, and use them with fresh media and reagents.

I have no viable cells after thawing my stock. What could be the cause of this?

Please see the possible causes and solutions we recommend below:

- Cells were stored incorrectly: Obtain a new stock and store in liquid nitrogen. Keep cells in liquid nitrogen until thawing.
- Homemade freezer stock is not viable: Freeze cells at a density recommended by the supplier. Use low-passage cells to make your own freezer stocks. Follow procedures for freezing cells exactly as recommended by the supplier. Obtain a new stock.
- Cells were thawed incorrectly: Follow procedures for thawing cells exactly as recommended by the supplier. Make sure that you thaw the frozen cells quickly but dilute them slowly using pre-warmed growth medium before plating.
- Thawing medium is not correct: Use the medium recommended by the supplier. Make sure the medium is pre-warmed.
- Cells are too dilute: Plate thawed cells at the highest density recommended by the supplier to optimize recovery.
- Cells not handled gently: Freezing and thawing procedures are stressful to most cells. Do not vortex, bang the flasks to dislodge the cells (except when culturing insect cells), or centrifuge cells at high speeds.
- Glycerol used in the freezing medium was stored in light (if applicable): If stored in light, glycerol gets converted into acrolein, which is toxic to cells. Obtain a new stock.

What factors can contribute to rapid cell death/culture failure?

There are a number of events that can contribute to this:

1. Incorrect CO2 levels-monitor the level of CO2 manually with a Fyrite kit, available from Bacharach (http://www.bacharach-inc.com/fyrite_analyzers.htm). Check if the manual readings concur with the readings displayed on the incubator. If the incubator has a trace readout, check the printout for fluctuations in CO2 level. Check the settings to insure that CO2 levels are set at appropriate levels for your cell line (usually between 5 and 10%). Check line connections frequently for leaks. Avoid frequent opening and closing of incubator doors.
2. Temperature fluctuations in the incubator-Monitor the temperature of incubator with a good thermometer inside the incubator.
3. Fungizone or other preventive antibiotics/antimycotics are present at toxic concentrations-use at recommended levels.
4. Humidity is incorrect-check the water level in the water pan. Humidity is vital to appropriate gas exchange for many types of cells and media (i.e., appropriate CO2 levels are largely irrelevant for most cultures if the humidity level is not high enough).
5. Incorrect osmotic pressure in medium-check osmolality of complete medium. Most mammalian cells can tolerate an osmolality of 260 to 350 mOsm/kg. Additions of reagents such as HEPES and drugs may affect osmolality.
6. Contamination by microorganisms-bacterial and fungal contaminations are usually easily visible; symptoms of mycoplasma contamination are more subtle, and careful monitoring of culture morphology and regular testing are necessary to detect this type of contamination.
7. Inappropriate medium is being used-double-check that the medium used is appropriate for your cell type and culture application. For example, ensure that the medium being used for serum-free culture is actually designed for serum-free culture; make sure that appropriate selective drugs are used at appropriate levels; check the expiration dates for the reagents being used; and store medium at appropriate temperatures in the dark.

Based on the sodium bicarbonate levels in the medium, what CO2 percentage is recommended?

If the media formulation contains:
- NaHCO3 (g/L) <1.5, it needs CO2 at 4%
- NaHCO3 (g/L) 1.5-2.2, it needs CO2 at 5%
- NaHCO3 (g/L) 2.2-3.4, it needs CO2 at 7%
- NaHCO3 (g/L) >3.5, it needs CO2 at 10%

However, there are some exceptions. Gibco DMEM has always been made according to Dulbecco's original published formulation, with 3.7 g/L sodium bicarbonate. Customers have been using this medium in CO2 incubators ranging from 5-10% CO2 for decades, usually with no trouble maintaining physiological pH; this also depends on the cell type. Once cells are growing, the pH will drop (due to metabolic accumulation of lactic acid).

I am happy with my present media, even though I am supplementing it with 10% FBS. Why would I change to Advanced media?

The Advanced media allow a reduction in serum usage from 50 to 90%. In the majority of cell lines, a 50% reduction can be achieved with no weaning procedure. Further reductions can be realized by weaning cells gradually. The amount of reduction in serum percentage beyond 50% will be cell line dependent. The advantages of using less serum include cost savings, particularly during those times when serum prices increase, extending the life of your serum lot, and reduced variability.

For more information on the Advanced DMEM and Advanced MEM, please search “Advanced Media” from our website home page.

How much serum should I add to my media?

The serum concentration will vary with the cell line and basal medium used. Please go here to see our recommended sera supplementations for tested cell lines (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/classical-media/advanced-d-mem-and-mem/recommended-sera-supplementations.html).

Do you offer assistance with media formulation development?

Yes. Our Gibco Custom Media Specialists and R&D staff can provide advice related to the addition or elimination of media components. For more comprehensive services, our PD-Direct Bioprocess Services team specializes in media development and optimization.

Who can answer questions related to the Gibco Media Configurator?

We have a dedicated team of Gibco Custom Media Specialists who can partner with you during customization, manufacturing, and delivery. Please email custommedia@lifetech.com or call the local phone number listed on www.thermofisher.com/mediaconfigurator with inquiries about your submission. For questions related to website effectiveness, please contact System Support through the toll-free or local phone number.

What are the benefits of using the Gibco Media Configurator?

Using the Gibco Media Configurator, you can view specific customization options for your cell culture product, online, at any time. Also, this tool provides faster pricing for custom media, generally within 48 business hours.

Can the Gibco Media Configurator be used for my own proprietary formulation?

No, the Gibco Media Configurator cannot be used to request pricing on non-Gibco cell culture products. However, we are very experienced at manufacturing customer-owned media formulations. Please visit www.thermofisher.com/custommedia or contact us at custommedia@lifetech.com for more information.

What aspects of cell culture media can be customized?

The Gibco Media Configurator allows you to add, remove, and adjust the concentration of components. In addition, you can select from a range of packaging, QC tests, and either cGMP or non-cGMP manufacturing.

Can any Gibco formulation be customized with the Gibco Media Configurator?

Almost all Gibco cell culture basal media products can be customized using the Gibco Media Configurator. To check on a particular product, use the Media Configurator page (https://www.thermofisher.com/us/en/home/life-science/bioproduction/custom-cell-culture-media-and-services/gibco-custom-media-configurator.html to search by Gibco catalog number, or browse our offering by description. For the customization of non-media Gibco products, such as buffers and growth factors, contact us at custommedia@lifetech.com.

Is it necessary to measure and adjust pH for the liquid media (classical or specialty) you sell, or are they already at the optimal pH?

No, you do not need to adjust the pH of the liquid media when you receive it. It should be in the correct pH range for use. After you add your supplements (if needed), you can check the pH again and adjust if necessary. However, usually it does not change significantly. It would be necessary to pH media that you make up yourself from a powder, with the exception of AGT media where pH and osmolality are auto-adjusted.

What quality water do you use in formulating your media?

Gibco cell culture media is formulated using water meeting all USP monograph requirements for Water For Injection.

How do you prepare cell culture media from powder or from concentrates?

You can find a protocol on our web site. In the main search box type “Media Preparation from Powder and Concentrates” and hit Enter or click Search. This should take you to a detailed protocol page.

How can I maintain an aseptic technique for cell culture?

Aseptic technique, designed to provide a barrier between the microrganisms in the environment and the sterile cell culture, depends upon a set of procedures to reduce the probability of contamination from these sources. The elements of aseptic technique are an aseptic work area, good personal hygiene, sterile (or sterile filtered) reagents and media, and aseptic handling. Go here to read more (https://www.thermofisher.com/us/en/home/references/gibco-cell-culture-basics/aseptic-technique.html) about aseptic technique or view our checklist (https://www.thermofisher.com/us/en/home/references/gibco-cell-culture-basics/aseptic-technique/aseptic-techniques-checklist.html).

What are the buffering conditions I should use for my CO2 incubator when culturing mammalian cells?

Most normal mammalian cell lines grow well at pH 7.4, and there is very little variability among different cell strains. While most researchers usually use 5-7% CO2 in air, 4-10% CO2 is common for most cell culture experiments. Read more about buffering conditions here (https://www.thermofisher.com/us/en/home/references/gibco-cell-culture-basics/cell-culture-environment/ph-CO2-levels.html).

Do you have a comparison of dishes, culture plates, and flasks by surface area, seeding density, cells at confluency, and volumes of Versene solution, trypsin, and growth medium?

Please see the comparison chart here (https://www.thermofisher.com/us/en/home/references/gibco-cell-culture-basics/cell-culture-protocols/cell-culture-useful-numbers.html).

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

Where can I find media formulations?

Please use our media formulation search tool (https://www.thermofisher.com/search/results?docTypes=MediaFormulation&persona=DocSupport&linkIn=true&query=).


WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.